Gene Transcripts

Gene Symbol
KIF25
Entrez Gene ID
3834
Gene Name
kinesin family member 25
Gene Aliases
KNSL3
Chromosome Location
Chr. 6: 168002543 - 168045106 on Build GRCh38
UniGene ID
Hs.150013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005355.3 NP_005346.3 2, 3 357
NM_030615.2 NP_085118.2 2, 3 357
XM_011535802.2 XP_011534104.1 4, 5 464
XM_011535803.2 XP_011534105.1 4, 5 464
GenBank mRNA AB012722.2 - 2, 3 357
AK289488.1 - 5 610
BC130489.1 - 3 405

View Full Product Details