Gene Transcripts

Gene Symbol
KIF25
Entrez Gene ID
3834
Gene Name
kinesin family member 25
Gene Aliases
KNSL3
Chromosome Location
Chr. 6: 168002543 - 168045106 on Build GRCh38
UniGene ID
Hs.150013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005355.3 NP_005346.3 5 621
NM_030615.2 NP_085118.2 5 621
XM_011535802.2 XP_011534104.1 7 728
XM_011535803.2 XP_011534105.1 7 728
GenBank mRNA AB012722.2 - 5 621
AK289488.1 - 7 874
BC130489.1 - 5 669

View Full Product Details