Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012213.2 | NP_036345.2 | 2 | 616 | |
GenBank mRNA | AF090834.1 | - | 2 | 596 | |
AF097832.2 | - | 2 | 534 | ||
AF153679.1 | - | 2 | 574 | ||
BC000286.1 | - | 2 | 568 | 3357140 | |
BC052592.1 | - | 2 | 616 | 6421051 |