Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr. 10: 87751512 - 87841345 on Build GRCh38
UniGene ID
Hs.435948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321967.1 NP_001308896.1 3, 4 332
NM_001321968.1 NP_001308897.1 3, 4 332
NM_001321969.1 NP_001308898.1 3, 4 332
NM_032810.3 NP_116199.2 3, 4 704
NR_135914.1 3, 4 332
XM_005270252.4 XP_005270309.1 3, 4 426
XM_011540302.1 XP_011538604.1 1, 2 97
XM_011540303.2 XP_011538605.1 3, 4 379
XM_017016847.1 XP_016872336.1 3, 4 573
XM_017016848.1 XP_016872337.1 3, 4 666
XM_017016849.1 XP_016872338.1 3, 4 455
XM_017016850.1 XP_016872339.1 3, 4 506
XM_017016851.1 XP_016872340.1 3, 4 379
GenBank mRNA AF361493.1 - 2, 3 538
AK075223.1 - 3, 4 633
AK304297.1 - 1, 2 95
AL834370.1 - 2, 3 117
BC063530.1 - 2, 3 163
BC073998.1 - 2, 3 275 6199813

View Full Product Details