Gene Transcripts

Gene Symbol
PSMB5
Entrez Gene ID
5693
Gene Name
proteasome subunit beta 5
Gene Aliases
LMPX, MB1, X
Chromosome Location
Chr. 14: 23016543 - 23035220 on Build GRCh38
UniGene ID
Hs.422990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144932.2 NP_001138404.1 1 429
NM_002797.4 NP_002788.1 1 429
XM_005267871.2 XP_005267928.1 1 424
GenBank mRNA AK225888.1 - 1 194
AK300714.1 - 1 174
AK311895.1 - 1 185
BC057840.1 - 1 184 4795732
BC107720.1 - 1 180 6738409
BX248299.1 - 1 161
BX538001.1 - 1 176
S74378.1 - 1 21

View Full Product Details