Gene Transcripts

Gene Symbol
SLC16A6
Entrez Gene ID
9120
Gene Name
solute carrier family 16 member 6
Gene Aliases
MCT6, MCT7
Chromosome Location
Chr. 17: 68267026 - 68291264 on Build GRCh38
UniGene ID
Hs.42645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174166.1 NP_001167637.1 3 434
NM_004694.4 NP_004685.2 2 307
XM_005257789.3 XP_005257846.2 2 472
XM_011525461.2 XP_011523763.1 2 151
XM_017025291.1 XP_016880780.1 3 271
XM_017025292.1 XP_016880781.1 2 334
GenBank mRNA AK290417.1 - 2 183
AK313121.1 - 2 173
AX750581.1 - 3 286
AX750582.1 - 1 121
BC064832.1 - 2 309 6498761
U79745.1 - 3 286

View Full Product Details