Gene Transcripts

Gene Symbol
SLC25A1
Entrez Gene ID
6576
Gene Name
solute carrier family 25 member 1
Gene Aliases
CTP, D2L2AD, SEA, SLC20A3
Chromosome Location
Chr. 22: 19175575 - 19178863 on Build GRCh38
UniGene ID
Hs.111024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256534.1 NP_001243463.1 6 876
NM_001287387.1 NP_001274316.1 7 730
NM_005984.4 NP_005975.1 7 895
NR_046298.2 6 756
GenBank mRNA AK292313.1 - 7 799
AK298204.1 - 7 728
BC004980.1 - 7 756 2957917
BC008061.2 - 7 758 2958945
BC018590.1 - 7 653
BI160240.1 - 6 621
HM037273.1 - 6 731
L75823.1 - 7 804
L77567.1 - 7 777
U25147.1 - 7 779

View Full Product Details