Gene Transcripts

Gene Symbol
UBE4A
Entrez Gene ID
9354
Gene Name
ubiquitination factor E4A
Gene Aliases
E4, UBOX2, UFD2
Chromosome Location
Chr. 11: 118359581 - 118399211 on Build GRCh38
UniGene ID
Hs.602967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204077.1 NP_001191006.1 20 5754
NM_004788.3 NP_004779.2 20 5775
GenBank mRNA D50916.1 - 20 5711

View Full Product Details