Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304485.1 | NP_001291414.1 | 5 | 925 | |
NM_001304486.1 | NP_001291415.1 | 4 | 803 | ||
NM_001304487.1 | NP_001291416.1 | 4 | 837 | ||
NM_001304488.1 | NP_001291417.1 | 5 | 822 | ||
NM_001304489.1 | NP_001291418.1 | 5 | 807 | ||
NM_001304490.1 | NP_001291419.1 | 5 | 804 | ||
NM_001304491.1 | NP_001291420.1 | 4 | 716 | ||
NM_012460.3 | NP_036592.1 | 6 | 1114 | ||
NR_130750.1 | 5 | 1049 | |||
NR_130751.1 | 4 | 739 | |||
NR_130752.1 | 3 | 738 | |||
NR_130753.1 | 3 | 730 | |||
GenBank mRNA | AF150100.1 | - | 5 | 805 | |
AK000666.1 | - | 5 | 823 | ||
BC020213.1 | - | 6 | 1014 | 4640518 | |
BC054875.1 | - | 4 | 727 | 6663143 | |
BG249379.1 | - | 5 | 693 |