Gene Transcripts

Gene Symbol
NPLOC4
Entrez Gene ID
55666
Gene Name
NPL4 homolog, ubiquitin recognition factor
Gene Aliases
NPL4
Chromosome Location
Chr. 17: 81556885 - 81637153 on Build GRCh38
UniGene ID
Hs.464333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017921.3 NP_060391.2 3, 4 441
XM_011524979.1 XP_011523281.1 3, 4 436
XM_011524980.1 XP_011523282.1 3, 4 436
XM_011524981.1 XP_011523283.1 3, 4 436
XM_011524982.2 XP_011523284.1 3, 4 436
GenBank mRNA AB040932.1 - 3, 4 348
AK000664.1 - 3, 4 350
AK023046.1 - 1, 2 77
AK023272.1 - 1 20
AK294473.1 - 3, 4 367
BC025930.1 - 3, 4 382 5017718
DC322260.1 - 3, 4 400
HY116973.1 - 3, 4 443

View Full Product Details