Gene Transcripts

Gene Symbol
FCN1
Entrez Gene ID
2219
Gene Name
ficolin 1
Gene Aliases
FCNM
Chromosome Location
Chr. 9: 134909585 - 134917960 on Build GRCh38
UniGene ID
Hs.440898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK314867.1 - 8, 9 774
BC020635.1 - 8, 9 756 4717731
D83920.1 - 8, 9 730

View Full Product Details