Gene Transcripts

Gene Symbol
PPP1CA
Entrez Gene ID
5499
Gene Name
protein phosphatase 1 catalytic subunit alpha
Gene Aliases
PP-1A, PP1A, PP1alpha, PPP1A
Chromosome Location
Chr. 11: 67398181 - 67401905 on Build GRCh38
UniGene ID
Hs.183994
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008709.1 NP_001008709.1 3 416
NM_002708.3 NP_002699.1 3 383
NM_206873.1 NP_996756.1 2 251
GenBank mRNA AK098311.1 - 5 1192
AK127616.1 - 4 600
AK313586.1 - 3 292
BC001888.1 - 3 304 3546588
BC004482.2 - 3 324 3689198
BC008010.1 - 3 324 3528742
BE278068.1 - 2 171
BE793459.1 - 3 386
BG480286.1 - 3 381
BM799599.1 - 2 190
BT006629.1 - 3 260
J04759.1 - 2 194
M63960.1 - 3 289
S57501.1 - 3 304
X70848.1 - 3 287

View Full Product Details