Gene Transcripts

Gene Symbol
PEF1
Entrez Gene ID
553115
Gene Name
penta-EF-hand domain containing 1
Gene Aliases
ABP32, PEF1A
Chromosome Location
Chr. 1: 31629862 - 31645237 on Build GRCh38
UniGene ID
Hs.470417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012392.3 NP_036524.1 5 1522
NR_033686.1 4 1221
NR_033688.1 5 1363
XM_011541745.1 XP_011540047.1 5 1149
XM_011541746.2 XP_011540048.1 5 1616
XM_011541747.1 XP_011540049.1 5 1491
XM_017001680.1 XP_016857169.1 5 1480
XM_017001681.1 XP_016857170.1 5 2787
GenBank mRNA AB018357.1 - 5 1161
AB026628.1 - 5 1161
AK001420.1 - 5 1174
AK092483.1 - 1 2516
AY359011.1 - 5 1165
BC002773.2 - 5 1151 3615612
BC012561.1 - 5 1167 4273213

View Full Product Details