Gene Transcripts

Gene Symbol
PRRC2C
Entrez Gene ID
23215
Gene Name
proline rich coiled-coil 2C
Gene Aliases
BAT2-iso, BAT2D1, BAT2L2, XTP2
Chromosome Location
Chr. 1: 171485496 - 171593511 on Build GRCh38
UniGene ID
Hs.494614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015172.3 NP_055987.2 12 1619
XM_005245015.2 XP_005245072.1 12 1656
XM_005245016.4 XP_005245073.1 12 1636
XM_005245017.2 XP_005245074.1 12 1656
XM_005245018.2 XP_005245075.1 12 1631
XM_005245019.2 XP_005245076.1 12 1656
XM_005245020.2 XP_005245077.1 12 1656
XM_005245023.2 XP_005245080.1 12 1656
XM_005245024.2 XP_005245081.1 12 1656
XM_005245025.2 XP_005245082.1 12 1656
XM_005245026.2 XP_005245083.1 12 1656
XM_005245027.2 XP_005245084.1 12 1656
XM_005245028.2 XP_005245085.1 12 1656
XM_005245029.2 XP_005245086.1 12 1656
XM_006711238.3 XP_006711301.1 12 1636
XM_017000760.1 XP_016856249.1 12 1621
XM_017000761.1 XP_016856250.1 12 1656
XM_017000762.1 XP_016856251.1 12 1656
GenBank mRNA AF253978.1 - 11 1424
AF488829.1 - 11 1353
AK298887.1 - 650
AL096857.1 - 11 1410
BC001260.1 - 12 1590
BC058930.1 - 12 1602
BC066968.1 - 12 1640

View Full Product Details