Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr. 1: 160343056 - 160358952 on Build GRCh38
UniGene ID
Hs.517249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290184.1 NP_001277113.1 10, 11 1372
NM_001290186.1 NP_001277115.1 6, 7 819
NM_015331.2 NP_056146.1 9, 10 1233
XM_005245053.4 XP_005245110.1 9, 10 1450
XM_011509363.2 XP_011507665.1 9, 10 1450
GenBank mRNA AF240468.1 - 9, 10 1251
AK296153.1 - 4, 5 518
AK299142.1 - 6, 7 706
AK314764.1 - 9, 10 1140
AY359120.1 - 9, 10 1110
BC047621.1 - 10, 11 1329 4827966
D87442.1 - 9, 10 1107

View Full Product Details