Gene Transcripts

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 7 774
GenBank mRNA AK293928.1 - 7 710
AK295146.1 - 7 774
BC099909.1 - 7 724

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256532.1 NP_001243461.1 7 728
NM_001256533.1 NP_001243462.1 7 728
NM_018116.3 NP_060586.2 7 728
NR_046292.1 7 850
NR_046293.1 7 792
NR_046294.1 5 714
NR_046295.1 6 652
XM_006711421.1 XP_006711484.1 6 687
XM_006711423.2 XP_006711486.1 7 736
XM_011509695.1 XP_011507997.1 7 686
XM_011509696.1 XP_011507998.1 7 686
XM_011509697.1 XP_011507999.1 7 686
XM_011509698.1 XP_011508000.1 7 686
XM_011509699.1 XP_011508001.1 7 686
XM_011509700.1 XP_011508002.1 7 686
XM_011509701.1 XP_011508003.1 7 686
XM_011509702.1 XP_011508004.1 7 686
XM_011509703.1 XP_011508005.1 6 1328
XM_011509704.1 XP_011508006.1 7 1251
XM_011509705.1 XP_011508007.1 6 1265
XM_011509706.1 XP_011508008.1 7 685
XM_011509707.1 XP_011508009.1 7 1193
XM_011509708.1 XP_011508010.1 7 1129
XM_011509709.2 XP_011508011.1 7 686
XM_017001603.1 XP_016857092.1 7 749
XM_017001604.1 XP_016857093.1 7 687
XM_017001605.1 XP_016857094.1 7 1218
XM_017001606.1 XP_016857095.1 7 1129
XM_017001607.1 XP_016857096.1 7 1267
XM_017001608.1 XP_016857097.1 6 714
XM_017001609.1 XP_016857098.1 7 736
XM_017001610.1 XP_016857099.1 7 724
XM_017001611.1 XP_016857100.1 7 1255
XM_017001612.1 XP_016857101.1 6 629
XM_017001613.1 XP_016857102.1 6 627
XM_017001614.1 XP_016857103.1 7 736
XM_017001615.1 XP_016857104.1 7 798
XM_017001616.1 XP_016857105.1 3 430
GenBank mRNA AF111708.1 - 4 417
AF272833.1 - 7 606
AK001366.1 - 7 657
AK056128.1 - 5 643
AK092950.1 - 6 552
AK222863.1 - 7 654
AK290610.1 - 7 703
AK297140.1 - 7 779
AK309573.1 - 7 713
AY334564.1 - 6 771
BC002535.2 - 7 673 3140498
BC070067.1 - 7 744
BX537684.1 - 6 604
CR749791.1 - 7 654
CR936872.1 - 3 1696

View Full Product Details