Gene Transcripts

Gene Symbol
PSMD1
Entrez Gene ID
5707
Gene Name
proteasome 26S subunit, non-ATPase 1
Gene Aliases
P112, Rpn2, S1
Chromosome Location
Chr. 2: 231056864 - 231172827 on Build GRCh38
UniGene ID
Hs.3887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191037.1 NP_001177966.1 17, 18 2168
NM_002807.3 NP_002798.2 17, 18 2168
NR_034059.1 16, 17 1998
GenBank mRNA AK307415.1 - 16, 17 1995
AK307513.1 - 15, 16 1870
AK312528.1 - 17, 18 2125
BC001053.2 - 17, 18 2125
BC014013.2 - 17, 18 2137
BC020223.1 - 17, 18 2115
BC032445.1 - 17, 18 2140
BC039845.1 - 17, 18 2091
BC047897.1 - 17, 18 2090
BC064398.1 - 17, 18 2139
BC073833.1 - 17, 18 2092
BC094720.1 - 17, 18 2154 4826340
BC112344.1 - 17, 18 2129 40008454
BC114434.1 - 17, 18 2129 40008444
D44466.1 - 17, 18 2086

View Full Product Details