Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168724.1 | NP_001162195.1 | 2 | 358 | |
NM_022918.3 | NP_075069.3 | 2 | 358 | ||
NR_033149.1 | 2 | 358 | |||
GenBank mRNA | AK291804.1 | - | 2 | 268 | |
AK302473.1 | - | 2 | 162 | ||
BC030952.1 | - | 2 | 217 | ||
BC051462.1 | - | 2 | 179 | 5278478 | |
BX648678.1 | - | 2 | 344 |