Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168724.1 | NP_001162195.1 | 14 | 1466 | |
NM_022918.3 | NP_075069.3 | 15 | 1532 | ||
NR_033149.1 | 14 | 1498 | |||
XM_017018140.1 | XP_016873629.1 | 14 | 1397 | ||
XM_017018141.1 | XP_016873630.1 | 13 | 1081 | ||
XM_017018142.1 | XP_016873631.1 | 13 | 1034 | ||
GenBank mRNA | AK000684.1 | - | 12 | 1075 | |
AK025757.1 | - | 13 | 1080 | ||
AK225364.1 | - | 13 | 1080 | ||
AK291804.1 | - | 14 | 1376 | ||
AK296774.1 | - | 11 | 1070 | ||
AK302473.1 | - | 14 | 1302 | ||
AL834493.1 | - | 6 | 519 | ||
BC012553.1 | - | 580 | |||
BC051462.1 | - | 15 | 1353 | 5278478 | |
BX648678.1 | - | 14 | 1452 |