Gene Transcripts

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
Chromosome Location
Chr. 1: 68098459 - 68233000 on Build GRCh38
UniGene ID
Hs.647659
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002292.3 NP_001002292.3 2 451
NM_024911.6 NP_079187.3 2 512
XM_011542191.2 XP_011540493.1 2 435
XM_011542192.2 XP_011540494.1 2 445
XM_017002390.1 XP_016857879.1 2 279
GenBank mRNA AB097018.1 - 2 434
AK074583.1 - 2 467
AK074984.1 - 2 434
AK309779.1 - 2 468
AY359035.1 - 2 494
BC007211.1 - 1 50
BC110826.1 - 2 413 6183768
BC137109.1 - 2 309 9021106
BC137113.1 - 2 309 9021110
BG701224.1 - 2 487
BX538320.1 - 2 451
DQ323735.1 - 2 481

View Full Product Details