Gene Transcripts

Gene Symbol
TSPAN18
Entrez Gene ID
90139
Gene Name
tetraspanin 18
Gene Aliases
TSPAN
Chromosome Location
Chr. 11: 44727148 - 44932427 on Build GRCh38
UniGene ID
Hs.385634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130783.4 NP_570139.3 7 840
XM_005253217.3 XP_005253274.1 9 1024
XM_006718372.3 XP_006718435.1 9 1111
XM_006718373.3 XP_006718436.1 8 1097
XM_011520459.2 XP_011518761.1 8 987
XM_017018530.1 XP_016874019.1 8 862
XM_017018531.1 XP_016874020.1 7 784
GenBank mRNA AK027715.1 - 6 649
AK075547.1 - 5 573
AY358087.1 - 6 763
BC019342.2 - 5 612 3357734

View Full Product Details