Gene Transcripts

Gene Symbol
EML3
Entrez Gene ID
256364
Gene Name
echinoderm microtubule associated protein like 3
Gene Aliases
ELP95
Chromosome Location
Chr. 11: 62602218 - 62613020 on Build GRCh38
UniGene ID
Hs.379785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300793.1 NP_001287722.1 13 1852
NM_001300794.1 NP_001287723.1 13 1849
NM_153265.2 NP_694997.2 13 1849
XM_005273878.3 XP_005273935.1 13 2107
XM_006718489.3 XP_006718552.1 13 2224
XM_006718490.3 XP_006718553.1 13 2224
XM_006718491.3 XP_006718554.1 13 2221
XM_011544896.2 XP_011543198.1 13 2221
XM_011544897.2 XP_011543199.1 12 2068
XM_017017480.1 XP_016872969.1 13 1595
XM_017017481.1 XP_016872970.1 12 1481
XM_017017482.1 XP_016872971.1 13 1595
XM_017017483.1 XP_016872972.1 13 1652
GenBank mRNA AK093146.1 - 12 1524
AK128679.1 - 12 2266
AK296901.1 - 13 1614
AK310579.1 - 15 1675
AL833388.1 - 10 1104
BC075824.1 - 12 1455
BC108280.1 - 13 1633 3915493

View Full Product Details