Gene Transcripts

Gene Symbol
FAM19A3
Entrez Gene ID
284467
Gene Name
family with sequence similarity 19 member A3, C-C motif chemokine like
Gene Aliases
TAFA-3, TAFA3
Chromosome Location
Chr. 1: 112719003 - 112727235 on Build GRCh38
UniGene ID
Hs.439116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004440.1 NP_001004440.1 5 588
NM_182759.2 NP_877436.1 5 520
XM_005270766.3 XP_005270823.1 6 807
XM_006710582.3 XP_006710645.1 6 875
GenBank mRNA AY325116.1 - 5 520
AY325119.1 - 5 588
BC112268.1 - 5 582 8327736
BC117479.1 - 5 582 40126030
BC143949.1 - 5 551
BC144042.1 - 5 571
BC144043.1 - 5 514 9052563

View Full Product Details