Gene Transcripts

Gene Symbol
CCDC84
Entrez Gene ID
338657
Gene Name
coiled-coil domain containing 84
Gene Aliases
DLNB14
Chromosome Location
Chr. 11: 118998067 - 119015793 on Build GRCh38
UniGene ID
Hs.534613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198489.2 NP_940891.1 8 767
NR_104049.1 8 795
NR_104050.1 8 795
XM_011542799.1 XP_011541101.1 8 795
XM_011542800.2 XP_011541102.1 6 475
XM_017017652.1 XP_016873141.1 8 3549
GenBank mRNA AB094093.1 - 8 758
AW014772.1 - 1 85
BC126469.1 - 8 732 8992185
BC126471.1 - 8 732 8992187
BC144209.1 - 8 760
BC144210.1 - 8 760
DB565963.1 - 1 22

View Full Product Details