Gene Transcripts

Gene Symbol
ELP6
Entrez Gene ID
54859
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C3orf75, TMEM103
Chromosome Location
Chr. 3: 47495640 - 47513711 on Build GRCh38
UniGene ID
Hs.311100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031703.2 NP_001026873.2 5 508
XM_005265241.3 XP_005265298.1 4 434
XM_011533851.2 XP_011532153.1 5 510
XM_011533852.2 XP_011532154.1 5 846
XM_011533853.2 XP_011532155.1 5 487
XM_011533854.2 XP_011532156.1 4 411
XM_011533856.2 XP_011532158.1 4 466
XM_011533857.2 XP_011532159.1 4 636
XM_017006669.1 XP_016862158.1 5 510
XM_017006670.1 XP_016862159.1 5 779
GenBank mRNA AK000218.1 - 5 493
AK225721.1 - 5 490
AK298202.1 - 4 443
BC000623.2 - 5 412 3346325
BC014204.2 - 5 412 4620013
DQ787088.1 - 5 389

View Full Product Details