Gene Transcripts

Gene Symbol
ASNSD1
Entrez Gene ID
54529
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
NBLA00058, NS3TP1
Chromosome Location
Chr. 2: 189661399 - 189670831 on Build GRCh38
UniGene ID
Hs.101364
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019048.2 NP_061921.1 4 536
XM_017004381.1 XP_016859870.1 3 267
XM_017004382.1 XP_016859871.1 3 1052
GenBank mRNA AK000759.1 - 4 515
AY116969.1 - 1 102
BC001243.1 - 4 441 3453136
DA771653.1 - 4 536

View Full Product Details