Gene Transcripts

Gene Symbol
IRGC
Entrez Gene ID
56269
Gene Name
immunity related GTPase cinema
Gene Aliases
CINEMA, IFGGE, IRGC1, Iigp5, R30953_1
Chromosome Location
Chr. 19: 43716062 - 43720021 on Build GRCh38
UniGene ID
Hs.515444
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019612.3 NP_062558.1 2 1575
GenBank mRNA BC066939.1 - 2 1551

View Full Product Details