Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320395.1 | NP_001307324.1 | 4 | 1283 | |
NM_001320397.1 | NP_001307326.1 | 5 | 1240 | ||
NM_001320398.1 | NP_001307327.1 | 5 | 1336 | ||
NM_001320399.1 | NP_001307328.1 | 6 | 1465 | ||
NM_001320400.1 | NP_001307329.1 | 3 | 1208 | ||
NM_001320401.1 | NP_001307330.1 | 4 | 1264 | ||
NM_020357.2 | NP_065090.1 | 5 | 1339 | ||
NR_135223.1 | 5 | 1287 | |||
NR_135224.1 | 4 | 1124 | |||
NR_135225.1 | 5 | 1284 | |||
NR_135226.1 | 3 | 1156 | |||
NR_135227.1 | 5 | 1540 | |||
NR_135228.1 | 4 | 1231 | |||
NR_135229.1 | 5 | 1468 | |||
NR_135230.1 | 4 | 1212 | |||
NR_135231.1 | 3 | 1049 | |||
NR_135232.1 | 6 | 1382 | |||
XM_017006922.1 | XP_016862411.1 | 4 | 1194 | ||
GenBank mRNA | AB037675.1 | - | 5 | 1235 | |
AK025551.1 | - | 5 | 1179 | ||
AK098760.1 | - | 5 | 1218 | ||
BC013916.1 | - | 4 | 1191 | 3929908 | |
BC022001.2 | - | 5 | 1237 | 4823990 | |
BC107796.1 | - | 5 | 1181 | ||
BX647886.1 | - | 1 | 1925 | ||
CR627371.1 | - | 4 | 1024 |