Gene Transcripts

Gene Symbol
PAQR3
Entrez Gene ID
152559
Gene Name
progestin and adipoQ receptor family member 3
Gene Aliases
RKTG
Chromosome Location
Chr. 4: 78887225 - 78939428 on Build GRCh38
UniGene ID
Hs.657312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040202.1 NP_001035292.1 6 3395
GenBank mRNA AK126273.1 - 3 2586
AL080110.1 - 1 1469
BC047510.1 - 7 3649 5297045
BX648346.1 - 5 3026

View Full Product Details