Gene Transcripts

Gene Symbol
CCDC36
Entrez Gene ID
339834
Gene Name
coiled-coil domain containing 36
Gene Aliases
CT74
Chromosome Location
Chr. 3: 49198428 - 49258104 on Build GRCh38
UniGene ID
Hs.631931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135197.1 NP_001128669.1 7, 8 731
NM_178173.3 NP_835467.2 9, 10 1029
XM_006713124.3 XP_006713187.1 7, 8 906
XM_011533670.2 XP_011531972.1 6, 7 740
XM_011533671.2 XP_011531973.1 6, 7 734
XM_011533672.2 XP_011531974.1 7, 8 1099
XM_011533673.2 XP_011531975.1 7, 8 1131
GenBank mRNA AK058049.1 - 9, 10 920
AK303033.1 - 7, 8 731
BC036202.1 - 9, 10 1029 4836976

View Full Product Details