Gene Transcripts

Gene Symbol
FAM19A4
Entrez Gene ID
151647
Gene Name
family with sequence similarity 19 member A4, C-C motif chemokine like
Gene Aliases
TAFA-4, TAFA4
Chromosome Location
Chr. 3: 68731764 - 68932610 on Build GRCh38
UniGene ID
Hs.187873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005527.2 NP_001005527.1 5, 6 830
NM_182522.4 NP_872328.1 5, 6 912
GenBank mRNA AK057890.1 - 4, 5 528
AY325117.1 - 5, 6 884
BC031566.1 - 5, 6 744 5195201

View Full Product Details