Gene Transcripts

Gene Symbol
LRP12
Entrez Gene ID
29967
Gene Name
LDL receptor related protein 12
Gene Aliases
ST7
Chromosome Location
Chr. 8: 104489231 - 104589024 on Build GRCh38
UniGene ID
Hs.600630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135703.2 NP_001129175.1 2 327
NM_013437.4 NP_038465.1 3 384
GenBank mRNA AB208957.1 - 1 59
AF064841.1 - 2 319
AF166350.1 - 3 299
AK289752.1 - 3 288
AK299374.1 - 2 324
AL698796.1 - 3 352
BC032109.2 - 3 260 4842797
CR749434.1 - 2 299
DC372924.1 - 2 324

View Full Product Details