Gene Transcripts

Gene Symbol
SIT1
Entrez Gene ID
27240
Gene Name
signaling threshold regulating transmembrane adaptor 1
Gene Aliases
SIT, SIT-R
Chromosome Location
Chr. 9: 35649298 - 35650950 on Build GRCh38
UniGene ID
Hs.88012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014450.2 NP_055265.1 5 1163
GenBank mRNA AJ010059.1 - 5 1154

View Full Product Details