Gene Transcripts
- Gene Symbol
- LYRM2
- Entrez Gene ID
- 57226
- Gene Name
- LYR motif containing 2
- Gene Aliases
- DJ122O8.2
- Chromosome Location
- Chr. 6: 89632224 - 89638755 on Build GRCh38
- UniGene ID
- Hs.177275
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020466.4 |
NP_065199.1
|
2, 3 |
233 |
|
NR_028494.1 |
|
2, 3 |
375 |
|
NR_028495.1 |
|
2, 3 |
243 |
|
GenBank mRNA |
AK024783.1 |
- |
2, 3 |
231 |
|
AK311180.1 |
- |
2, 3 |
292 |
|
AK311948.1 |
- |
2, 3 |
218 |
|
AV710669.1 |
- |
2, 3 |
233 |
|
BC009782.1 |
- |
2, 3 |
227 |
4092710 |
BC104668.1 |
- |
2, 3 |
216 |
6717725 |
BP280644.1 |
- |
2, 3 |
233 |
|
View Full Product Details