Gene Transcripts

Gene Symbol
FAM71F1
Entrez Gene ID
84691
Gene Name
family with sequence similarity 71 member F1
Gene Aliases
FAM137A, NYD-SP18
Chromosome Location
Chr. 7: 128709061 - 128731743 on Build GRCh38
UniGene ID
Hs.732440
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282788.1 NP_001269717.1 3 719
NM_001282789.1 NP_001269718.1 4 666
NM_032599.3 NP_115988.1 3 719
NR_104242.1 3 719
NR_104243.1 4 608
XM_017012743.1 XP_016868232.1 3 762
GenBank mRNA AF367470.1 - 3 672
AK093161.1 - 4 663
AK302221.1 - 4 507
AK310308.1 - 3 666
AK310426.1 - 4 575
AK313608.1 - 3 670
BC037248.1 - 3 658 5165912

View Full Product Details