Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152274.4 | NP_689487.2 | 5 | 781 | |
XM_005277920.4 | XP_005277977.1 | 5 | 739 | ||
XM_011531214.2 | XP_011529516.1 | 5 | 1305 | ||
XM_011531215.2 | XP_011529517.1 | 5 | 1171 | ||
GenBank mRNA | BC001909.1 | - | 5 | 721 | |
BC007232.1 | - | 5 | 721 | 3347302 | |
BC032121.2 | - | 5 | 729 | 4575331 | |
BC071851.1 | - | 5 | 582 | 5934127 |