Gene Transcripts

Gene Symbol
RWDD4
Entrez Gene ID
201965
Gene Name
RWD domain containing 4
Gene Aliases
FAM28A, RWDD4A
Chromosome Location
Chr. 4: 183639635 - 183659219 on Build GRCh38
UniGene ID
Hs.133337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307922.1 NP_001294851.1 4 539
NM_152682.3 NP_689895.2 4 543
GenBank mRNA AK021702.1 - 4 467
AK293274.1 - 2 284
AK315515.1 - 4 477
AL540937.3 - 4 480
AL559547.3 - 4 476
BC017472.1 - 4 478 3909581
BC067752.1 - 4 502 5270461
BC107432.1 - 4 324 6104330

View Full Product Details