Gene Transcripts

Gene Symbol
SRRM3
Entrez Gene ID
222183
Gene Name
serine/arginine repetitive matrix 3
Gene Aliases
Chromosome Location
Chr. 7: 76201893 - 76287292 on Build GRCh38
UniGene ID
Hs.511025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110199.2 NP_001103669.1 15 2952
NM_001291831.1 NP_001278760.1 16 3031
GenBank mRNA AK092590.1 - 3 1370
AK094397.1 - 1 1563
BC063788.1 - 1 1291
BC096755.1 - 4 1481

View Full Product Details