Gene Transcripts

Gene Symbol
C8orf31
Entrez Gene ID
286122
Gene Name
chromosome 8 open reading frame 31
Gene Aliases
Chromosome Location
Chr. 8: 143038983 - 143054303 on Build GRCh38
UniGene ID
Hs.660382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173687.3 NP_775958.1 5, 6 847
NR_126398.1 7, 8 1029
NR_126399.1 5, 6 838
GenBank mRNA AK094450.1 - 5, 6 795
BC063497.1 - 5, 6 749
BC073830.1 - 7, 8 977
BC120993.2 - 5, 6 773

View Full Product Details