Gene Transcripts

Gene Symbol
TMC1
Entrez Gene ID
117531
Gene Name
transmembrane channel like 1
Gene Aliases
DFNA36, DFNB11, DFNB7
Chromosome Location
Chr. 9: 72521801 - 72836351 on Build GRCh38
UniGene ID
Hs.670211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138691.2 NP_619636.2 24 3126
XM_017014256.1 XP_016869745.1 21 2849
GenBank mRNA AF417578.1 - 24 3126
AK098607.1 - 11 1484

View Full Product Details