Gene Transcripts

Gene Symbol
THAP6
Entrez Gene ID
152815
Gene Name
THAP domain containing 6
Gene Aliases
Chromosome Location
Chr. 4: 75513951 - 75547334 on Build GRCh38
UniGene ID
Hs.479971
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317791.1 NP_001304720.1 4 553
NM_144721.5 NP_653322.1 5 679
XM_005262772.2 XP_005262829.1 4 574
XM_011531666.2 XP_011529968.1 5 1116
XM_011531667.2 XP_011529969.1 5 639
XM_017007800.1 XP_016863289.1 4 719
GenBank mRNA AK313765.1 - 5 1063
AL833350.1 - 5 639
BC022989.2 - 4 593 5113206
BX647579.1 - 4 513

View Full Product Details