Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146256.1 | NP_001139728.1 | 3 | 807 | |
NM_001146257.1 | NP_001139729.1 | 3 | 807 | ||
NM_144969.2 | NP_659406.1 | 4 | 834 | ||
XM_006724624.3 | XP_006724687.1 | 4 | 371 | ||
XM_017029296.1 | XP_016884785.1 | 4 | 371 | ||
GenBank mRNA | AK056374.1 | - | 4 | 372 | |
AK122885.1 | - | 3 | 556 | ||
AK128587.1 | - | 3 | 342 | ||
AK303896.1 | - | 4 | 372 | ||
AY358786.1 | - | 3 | 728 | ||
BC103980.1 | - | 4 | 372 | 40016292 | |
BC103981.1 | - | 4 | 372 | 40016293 | |
BC103982.1 | - | 4 | 372 | 40016294 |