Gene Transcripts

Gene Symbol
SLITRK4
Entrez Gene ID
139065
Gene Name
SLIT and NTRK like family member 4
Gene Aliases
Chromosome Location
Chr. X: 143622790 - 143636107 on Build GRCh38
UniGene ID
Hs.272284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184749.2 NP_001171678.1 2 2732
NM_001184750.2 NP_001171679.1 2 2535
NM_173078.4 NP_775101.1 2 2685
XM_005262365.4 XP_005262422.1 2 2485
XM_011531265.2 XP_011529567.1 2 2530
GenBank mRNA AK057619.1 - 1 1402
AK092016.1 - 2 2400
AK315044.1 - 2 2535
AL713693.1 - 1 1865
BC040986.1 - 2 2694 4811695

View Full Product Details