Gene Transcripts

Gene Symbol
FAM83C
Entrez Gene ID
128876
Gene Name
family with sequence similarity 83 member C
Gene Aliases
C20orf128, dJ614O4.7
Chromosome Location
Chr. 20: 35285731 - 35292422 on Build GRCh38
UniGene ID
Hs.592149
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178468.5 NP_848563.1 4 2034
GenBank mRNA AK091116.1 - 5 1601
AK091138.1 - 3 1377
BC113483.1 - 4 2013 8322535

View Full Product Details