Gene Transcripts

Gene Symbol
PLEKHA3
Entrez Gene ID
65977
Gene Name
pleckstrin homology domain containing A3
Gene Aliases
FAPP1
Chromosome Location
Chr. 2: 178480410 - 178516324 on Build GRCh38
UniGene ID
Hs.41086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019091.3 NP_061964.3 6, 7 1065
XM_011511689.2 XP_011509991.1 7, 8 1174
GenBank mRNA AF286162.1 - 6, 7 904
AK000074.1 - 4, 5 468
AK074209.1 - 6, 7 1040
AK314459.1 - 6, 7 1069
BC044567.1 - 6, 7 759 5262097
DQ786259.1 - 3, 4 382
DQ786318.1 - 2, 3 222

View Full Product Details