Gene Transcripts

Gene Symbol
GGCT
Entrez Gene ID
79017
Gene Name
gamma-glutamylcyclotransferase
Gene Aliases
C7orf24, CRF21, GCTG, GGC
Chromosome Location
Chr. 7: 30496621 - 30504841 on Build GRCh38
UniGene ID
Hs.530024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199815.1 NP_001186744.1 4 844
NM_001199816.1 NP_001186745.1 3 688
NM_001199817.1 NP_001186746.1 2 542
NM_024051.3 NP_076956.1 4 824
NR_037669.1 3 678
GenBank mRNA AK021779.1 - 3 1303
BC000625.2 - 4 819 3346822
BC005356.1 - 3 676 4102029
BC013937.1 - 4 785 3997629
BC019243.1 - 4 816 3828006
BE885397.1 - 2 502
BG527593.1 - 3 667

View Full Product Details