Gene Transcripts

Gene Symbol
TMC7
Entrez Gene ID
79905
Gene Name
transmembrane channel like 7
Gene Aliases
Chromosome Location
Chr. 16: 18983934 - 19063942 on Build GRCh38
UniGene ID
Hs.187377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160364.2 NP_001153836.1 2 445
NM_001300732.1 NP_001287661.1 2 347
NM_001324263.1 NP_001311192.1 2 347
NM_001324265.1 NP_001311194.1 2 347
NM_001324268.1 NP_001311197.1 2 347
NM_024847.4 NP_079123.3 2 347
NR_136733.1 2 347
GenBank mRNA AK094664.1 - 2 347
AK293874.1 - 2 258
AY236498.1 - 2 281
AY263165.1 - 2 276
BC036205.1 - 2 276 5261157
BC047719.1 - 2 278 5743899
CR933669.1 - 2 445
DA161026.1 - 2 348

View Full Product Details