Gene Transcripts
- Gene Symbol
- SLC2A11
- Entrez Gene ID
- 66035
- Gene Name
- solute carrier family 2 member 11
- Gene Aliases
- GLUT10, GLUT11
- Chromosome Location
- Chr. 22: 23856703 - 23886112 on Build GRCh38
- UniGene ID
- Hs.661910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001024938.3 |
NP_001020109.1
|
8, 9 |
1247 |
|
NM_001024939.3 |
NP_001020110.1
|
8, 9 |
1135 |
|
NM_030807.4 |
NP_110434.3
|
9, 10 |
1358 |
|
NR_104247.1 |
|
7, 8 |
1094 |
|
GenBank mRNA |
AF443201.1 |
- |
9, 10 |
1189 |
|
AJ271290.1 |
- |
8, 9 |
1078 |
|
AK075467.1 |
- |
8, 9 |
1023 |
|
AK296169.1 |
- |
7, 8 |
863 |
|
AK308602.1 |
- |
9, 10 |
1152 |
|
AK314502.1 |
- |
9, 10 |
1129 |
|
BC094735.1 |
- |
8, 9 |
1064 |
5304025 |
BC100807.2 |
- |
7, 8 |
844 |
|
BC100808.2 |
- |
8, 9 |
993 |
40000757 |
BC100809.1 |
- |
8, 9 |
993 |
40000760 |
BC100810.1 |
- |
7, 8 |
840 |
|
CR456373.1 |
- |
8, 9 |
1024 |
|
View Full Product Details