Gene Transcripts

Gene Symbol
C9orf64
Entrez Gene ID
84267
Gene Name
chromosome 9 open reading frame 64
Gene Aliases
Chromosome Location
Chr. 9: 83938311 - 83956986 on Build GRCh38
UniGene ID
Hs.208914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317997.1 NP_001304926.1 5 1493
NM_032307.4 NP_115683.3 4 1891
GenBank mRNA AK090882.1 - 5 1493
BC004407.1 - 4 1866

View Full Product Details