Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290325.1 | NP_001277254.1 | 1 | 1063 | |
NM_001290326.1 | NP_001277255.1 | 1 | 1063 | ||
NM_032557.6 | NP_115946.2 | 1 | 1063 | ||
XM_011532360.2 | XP_011530662.1 | 1 | 551 | ||
GenBank mRNA | AK093754.1 | - | 1 | 286 | |
AK126943.1 | - | 2 | 891 | ||
AL833976.1 | - | 1 | 1035 | ||
BC039115.1 | - | 1 | 1063 | 5258804 | |
BC068975.1 | - | 1 | 1010 | 6067501 |